APO455829840 Apollo Scientific Ltd

3-Chloroisoquinoline-6-carboxaldehyde

  • MDL number MFCD22380179
  • Cas Number : 1337880-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Chloro-6-formylisoquinoline, 3-Chloro-6-formyl-2-azanaphthalene
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54530

Safety Information

signalword Warning
pcodes P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54530
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart