APO455829849 Apollo Scientific Ltd

3-Bromo-5-chloroisoquinoline

  • MDL number MFCD18793406
  • Cas Number : 1276056-67-7
  • Molecular Weight : 242.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-5-chloroisoquinoline
Inchi Code 1S/C9H5BrClN/c10-9-4-7-6(5-12-9)2-1-3-8(7)11/h1-5H
Inchi Key ILNONYMTPPUBOV-UHFFFAOYSA-N

Description

Synonym 3-Bromo-5-chloro-2-azanaphthalene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-chloroisoquinoline

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-chloroisoquinoline
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart