APO455829853 Apollo Scientific Ltd

4-Amino-3-chloroisoquinoline

  • MDL number MFCD08235154
  • Cas Number : 342899-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Chloroisoquinolin-4-amine, 4-Amino-3-chloro-2-azanaphthalene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-amino-3-chloroisoquinoline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-3-chloroisoquinoline
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart