APO455829863 Apollo Scientific Ltd

6-Amino-3-chloroisoquinoline

  • MDL number MFCD22380488
  • Cas Number : 1374652-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Chloroisoquinolin-6-amine, 6-Amino-3-chloro-2-azanaphthalene
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54566

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54566
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart