APO455829875 Apollo Scientific Ltd

3-Methoxyisoquinoline-7-boronic acid

  • MDL number MFCD19982319
  • Cas Number : 1645213-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 7-Borono-3-methoxyisoquinoline, 7-Borono-3-methoxy-2-azanaphthalene
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54599

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54599
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart