APO455829878 Apollo Scientific Ltd

Ethyl 1H-imidazole-2-carboxylate

  • MDL number MFCD03426031
  • Cas Number : 33543-78-1
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176-180
Synonym 2-(Ethoxycarbonyl)-1H-imidazole
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 48-51/0.3mm
Product Link https://store.apolloscientific.co.uk/product/ethyl-1h-imidazole-2-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-1h-imidazole-2-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart