APO455829879 Apollo Scientific Ltd

Ethyl 1-(tert-butyl)-5-methyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD01313625
  • Cas Number : 519056-54-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(tert-Butyl)-3-(ethoxycarbonyl)-5-methyl-1H-pyrazole
Purity 95
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 130/15mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=26210

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=26210
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart