APO455829880 Apollo Scientific Ltd

Ethyl 3-hydroxy-1H-pyrazole-4-carboxylate

  • MDL number MFCD12131138
  • Cas Number : 7251-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(Ethoxycarbonyl)-3-hydroxy-1H-pyrazole
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=51537

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=51537
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart