APO455829886 Apollo Scientific Ltd

4-(Methylsulphonyl)phenacyl bromide

  • MDL number MFCD00673134
  • Cas Number : 50413-24-6
  • Molecular Weight : 277.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 125-130
Iupac Name 2-bromo-1-[4-(methylsulfonyl)phenyl]ethanone
Inchi Code 1S/C9H9BrO3S/c1-14(12,13)8-4-2-7(3-5-8)9(11)6-10/h2-5H,6H2,1H3
Inchi Key JOCMYOUZIDSYFO-UHFFFAOYSA-N

Description

Synonym 2-Bromo-4'-(methylsulphonyl)acetophenone,2-Bromo-1-[4-(methylsulphonyl)phenyl]-1-ethanone
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 125-130
Product Link https://store.apolloscientific.co.uk/product/4-methylsulphonylphenacyl-bromide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-methylsulphonylphenacyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart