APO455829888 Apollo Scientific Ltd

Ethyl 5-methyl-1H-pyrazole-4-carboxylate

  • MDL number MFCD08443874
  • Cas Number : 85290-78-4
  • Molecular Weight : 154.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 49-55
Iupac Name ethyl 5-methyl-1H-pyrazole-4-carboxylate
Inchi Code 1S/C7H10N2O2/c1-3-11-7(10)6-4-8-9-5(6)2/h4H,3H2,1-2H3,(H,8,9)
Inchi Key HHYVTIKYZUMDIL-UHFFFAOYSA-N

Description

Synonym 4-(Ethoxycarbonyl)-5-methyl-1H-pyrazole, Ethyl 3-methyl-1H-pyrazole-4-carboxylate, 4-(Ethoxycarbonyl)-3-methyl-1H-pyrazole
Storage temp. Ambient Temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 49-55
Product Link https://store.apolloscientific.co.uk/product/ethyl-5-methyl-1h-pyrazole-4-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-5-methyl-1h-pyrazole-4-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart