APO455829892 Apollo Scientific Ltd

3,5-Dichlorobenzylamine

  • MDL number MFCD00052681
  • Cas Number : 39989-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (3,5-Dichlorophenyl)methylamine
Purity 98
Density 1.32
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 141-144/15mm
Product Link https://store.apolloscientific.co.uk/product/35-dichlorobenzylamine

Safety Information

signalword Danger
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/35-dichlorobenzylamine
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart