APO455829899 Apollo Scientific Ltd

1,3-Dimethyl-1H-imidazol-3-ium iodide

  • MDL number MFCD00159685
  • Cas Number : 4333-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=51715

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=51715
pictograms GHS07
hstatements H302+H312+H332;H413
Availability details for
Quantity
  • Availability
  • Add to Cart