APO455829901 Apollo Scientific Ltd

1H-Pyrazole-4-carboxaldehyde

  • MDL number MFCD02179514
  • Cas Number : 35344-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82-83
Synonym 4-Formyl-1H-pyrazole
Flash Point >110
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=51738

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=51738
pictograms GHS05;GHS07;GHS08
hstatements H302;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart