APO455829906 Apollo Scientific Ltd

3-Aminocyclohexan-1-one, N-BOC protected

  • MDL number MFCD03790905
  • Cas Number : 885280-38-6
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-oxocyclohexylcarbamate
Inchi Code 1S/C11H19NO3/c1-11(2,3)15-10(14)12-8-5-4-6-9(13)7-8/h8H,4-7H2,1-3H3,(H,12,14)
Inchi Key VGDCXKATFLOEHF-UHFFFAOYSA-N

Description

Synonym tert-Butyl (3-oxocyclohexyl)carbamate, 3-Oxocyclohexan-1-amine, N-BOC protected
Purity 97%
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-aminocyclohexan-1-one-n-boc-protected

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-aminocyclohexan-1-one-n-boc-protected
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart