APO455829909 Apollo Scientific Ltd

Methyl 4-amino-1,3-thiazole-5-carboxylate

  • MDL number MFCD09033876
  • Cas Number : 278183-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Amino-5-(methoxycarbonyl)-1,3-thiazole
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/methyl-4-amino-13-thiazole-5-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-amino-13-thiazole-5-carboxylate
pictograms GHS07
hstatements H315;H319;H412
Availability details for
Quantity
  • Availability
  • Add to Cart