APO455829914 Apollo Scientific Ltd

Morpholin-3-one

  • MDL number MFCD00631009
  • Cas Number : 109-11-5
  • Molecular Weight : 101.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-107
Synonym 3-Oxomorpholine
Physical Form Solid-Crystals
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/morpholin-3-one

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/morpholin-3-one
pictograms GHS05;GHS07
hstatements H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart