APO455829916 Apollo Scientific Ltd

3,5-Dichloro-2-iodotoluene

  • MDL number MFCD00052856
  • Cas Number : 175277-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43-45
Synonym 1,5-Dichloro-2-iodo-3-methylbenzene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=26289

Safety Information

signalword Warning
pcodes P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=26289
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart