APO455829917 Apollo Scientific Ltd

3-(Morpholin-4-yl)phenol

  • MDL number MFCD00051675
  • Cas Number : 27292-49-5
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 132-134
Iupac Name 3-(4-morpholinyl)phenol
Inchi Code 1S/C10H13NO2/c12-10-3-1-2-9(8-10)11-4-6-13-7-5-11/h1-3,8,12H,4-7H2
Inchi Key BMGSGGYIUOQZBZ-UHFFFAOYSA-N

Description

Synonym 4-(3-Hydroxyphenyl)morpholine
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 132-134
Product Link https://store.apolloscientific.co.uk/product/3-morpholin-4-ylphenol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-morpholin-4-ylphenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart