APO455829919 Apollo Scientific Ltd

5-Chloro-2-methoxybenzonitrile

  • MDL number MFCD00052867
  • Cas Number : 55877-79-7
  • Molecular Weight : 167.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 98-102
Iupac Name 5-chloro-2-methoxybenzonitrile
Inchi Code 1S/C8H6ClNO/c1-11-8-3-2-7(9)4-6(8)5-10/h2-4H,1H3
Inchi Key LREABOKKLIVXNA-UHFFFAOYSA-N

Description

Synonym 4-Chloro-2-cyanoanisole
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 98-102
Product Link https://store.apolloscientific.co.uk/product/5-chloro-2-methoxybenzonitrile

Safety Information

signalword Danger
pcodes P312;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-2-methoxybenzonitrile
pictograms GHS06
hstatements H301;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart