APO455829923 Apollo Scientific Ltd

3-Amino-7-bromoisoquinoline

  • MDL number MFCD11040207
  • Cas Number : 1192815-02-3
  • Molecular Weight : 223.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-bromo-3-isoquinolinamine
Inchi Code 1S/C9H7BrN2/c10-8-2-1-6-4-9(11)12-5-7(6)3-8/h1-5H,(H2,11,12)
Inchi Key FBTKUAFGTDENLW-UHFFFAOYSA-N

Description

Synonym 7-Bromoisoquinolin-3-amine, 3-Amino-7-bromo-2-azanaphthalene
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-7-bromoisoquinoline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-7-bromoisoquinoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart