APO455829930 Apollo Scientific Ltd

Ethyl 5,5-dimethyl-2,4-dioxohexanoate

  • MDL number MFCD00052319
  • Cas Number : 13395-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ethyl (trimethylacetyl)pyruvate, Ethyl 5,5-dimethyl-2,4-dioxocaproate
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 68-70/0.5mm
Product Link https://store.apolloscientific.co.uk/product/ethyl-55-dimethyl-24-dioxohexanoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-55-dimethyl-24-dioxohexanoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart