APO455829934 Apollo Scientific Ltd

4-Chloro-5-nitrophthalimide

  • MDL number MFCD00052331
  • Cas Number : 6015-57-2
  • Molecular Weight : 226.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 201-203
Iupac Name 5-chloro-6-nitro-1H-isoindole-1,3(2H)-dione
density 1.725
Inchi Code 1S/C8H3ClN2O4/c9-5-1-3-4(2-6(5)11(14)15)8(13)10-7(3)12/h1-2H,(H,10,12,13)
Inchi Key ADLVDYMTBOSDFE-UHFFFAOYSA-N

Description

Synonym 5-Chloro-6-nitroisoindolin-1,3-dione, 5-Chloro-6-nitro-1H-isoindole-1,3(2H)-dione
Density 1.725
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 201-203
Product Link https://store.apolloscientific.co.uk/product/4-chloro-5-nitrophthalimide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-5-nitrophthalimide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart