APO455829940 Apollo Scientific Ltd

3-Methylbenzylthiol

  • MDL number MFCD00022075
  • Cas Number : 25697-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Methylbenzyl mercaptan, (3-Methylphenyl)methanethiol
Purity 95
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 102-104/15mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=26342

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=26342
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart