APO455829941 Apollo Scientific Ltd

Cyclopentyl isothiocyanate

  • MDL number MFCD00040877
  • Cas Number : 33522-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Isothiocyanatocyclopentane
Purity 97
Density 1.03
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 75-76/10mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=26348

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=26348
pictograms GHS06
hstatements H301+H311+H331;H314
Availability details for
Quantity
  • Availability
  • Add to Cart