APO455829947 Apollo Scientific Ltd

2-Amino-4-methylbenzoic acid

  • MDL number MFCD00047853
  • Cas Number : 2305-36-4
  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 174-176
Iupac Name 2-amino-4-methylbenzoic acid
Inchi Code 1S/C8H9NO2/c1-5-2-3-6(8(10)11)7(9)4-5/h2-4H,9H2,1H3,(H,10,11)
Inchi Key RPGKFFKUTVJVPY-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 174-176
Product Link https://store.apolloscientific.co.uk/product/2-amino-4-methylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-4-methylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart