APO455829955 Apollo Scientific Ltd

1-(2,3-Dihydro-1H-inden-5-yl)ethan-1-one

  • MDL number MFCD00021246
  • Cas Number : 4228-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Acetylindane, (Indan-5-yl)ethan-1-one
Density 1.06
Storage temp. Ambient temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 85-87/0.5mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=26420

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=26420
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart