APO455829957 Apollo Scientific Ltd

5-Amino-4-bromo-3-phenyl-1H-pyrazole

  • MDL number MFCD00067992
  • Cas Number : 2845-78-5
  • Molecular Weight : 238.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 112-120
Iupac Name 4-bromo-3-phenyl-1H-pyrazol-5-amine
Inchi Code 1S/C9H8BrN3/c10-7-8(12-13-9(7)11)6-4-2-1-3-5-6/h1-5H,(H3,11,12,13)
Inchi Key QTNVXMOPTHGCII-UHFFFAOYSA-N

Description

Synonym 4-Bromo-3-phenyl-1H-pyrazol-5-amine, (5-Amino-4-bromo-1H-pyrazol-3-yl)benzene
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 112-120
Product Link https://store.apolloscientific.co.uk/product/5-amino-4-bromo-3-phenyl-1h-pyrazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-4-bromo-3-phenyl-1h-pyrazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart