APO455829963 Apollo Scientific Ltd

4-Ethylbenzhydrazide

  • MDL number MFCD00221478
  • Cas Number : 64328-55-8
  • Molecular Weight : 164.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 89-90
Iupac Name 4-ethylbenzohydrazide
Inchi Code 1S/C9H12N2O/c1-2-7-3-5-8(6-4-7)9(12)11-10/h3-6H,2,10H2,1H3,(H,11,12)
Inchi Key CBPNMGZKCDVJAE-UHFFFAOYSA-N

Description

Synonym 4-Ethylbenzoic acid hydrazide, 4-Ethylbenzohydrazide
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 89-90
Product Link https://store.apolloscientific.co.uk/product/4-ethylbenzhydrazide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-ethylbenzhydrazide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart