APO455829972 Apollo Scientific Ltd

S,S-Dimethylsulphoximine

  • MDL number MFCD02656633
  • Cas Number : 1520-31-6
  • Molecular Weight : 93.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 51-58
Iupac Name methyl(methylene)oxido-lambda~4~-sulfanylamine
density 1.182
Inchi Code 1S/C2H7NOS/c1-5(2,3)4/h1H2,2H3,(H2,3,4)
Inchi Key WRXZOSOVSAKERF-UHFFFAOYSA-N

Description

Synonym (S-Methylsulphonimidoyl)methane
Purity 98%
Density 1.182
Flash Point 100-102/1.5mm
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 51-58
Product Link https://store.apolloscientific.co.uk/product/ss-dimethylsulphoximine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ss-dimethylsulphoximine
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart