APO455829973 Apollo Scientific Ltd

5-Methyl-3-phenylisoxazol-4-amine

  • MDL number MFCD02677726
  • Cas Number : 21169-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51-53
Synonym 4-Amino-5-methyl-3-phenylisoxazole, 5-Methyl-3-phenyl-1,2-oxazol-4-amine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 131-133/0.5mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=26466

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=26466
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart