APO455829977 Apollo Scientific Ltd

5-Amino-3-(tert-butyl)-1-methyl-1H-pyrazole

  • MDL number MFCD00068002
  • Cas Number : 118430-73-2
  • Molecular Weight : 153.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(tert-Butyl)-1-methyl-1H-pyrazol-5-amine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-amino-3-tert-butyl-1-methyl-1h-pyrazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-3-tert-butyl-1-methyl-1h-pyrazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart