APO455829978 Apollo Scientific Ltd

4-(Benzylthio)-3-nitrobenzaldehyde

  • MDL number MFCD00173710
  • Cas Number : 175278-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-172
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=26491

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=26491
pictograms GHS07
hstatements H302+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart