APO455829979 Apollo Scientific Ltd

5-Amino-1-methylpyrazole-4-carboxamide

  • MDL number MFCD00111808
  • Cas Number : 18213-75-7
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 221-225
Iupac Name 5-amino-1-methyl-1H-pyrazole-4-carboxamide
Inchi Code 1S/C5H8N4O/c1-9-4(6)3(2-8-9)5(7)10/h2H,6H2,1H3,(H2,7,10)
Inchi Key JGSQVTVXGXOSCH-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 221-225
Product Link https://store.apolloscientific.co.uk/product/5-amino-1-methylpyrazole-4-carboxamide

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-1-methylpyrazole-4-carboxamide
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart