APO455829983 Apollo Scientific Ltd

1-Methyl-3-phenyl-1H-pyrazol-5-amine

  • MDL number MFCD00067874
  • Cas Number : 10199-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Amino-1-methyl-3-phenyl-1H-pyrazole, (5-Amino-1-methyl-1H-pyrazol-3-yl)benzene
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=12718

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=12718
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart