APO455829985 Apollo Scientific Ltd

Diethyl 2-(2-nitrobenzyl)malonate

  • MDL number MFCD00110674
  • Cas Number : 59803-35-9
  • Molecular Weight : 295.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name diethyl 2-(2-nitrobenzyl)malonate
Inchi Code 1S/C14H17NO6/c1-3-20-13(16)11(14(17)21-4-2)9-10-7-5-6-8-12(10)15(18)19/h5-8,11H,3-4,9H2,1-2H3
Inchi Key XKWHIUXRQYXRSG-UHFFFAOYSA-N

Description

Synonym Diethyl 2-(2-nitrobenzyl)propane-1,3-dioate
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/diethyl-2-2-nitrobenzylmalonate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/diethyl-2-2-nitrobenzylmalonate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart