APO455830001 Apollo Scientific Ltd

6-Bromo-3,4-dihydroquinolin-2(1H)-one

  • MDL number MFCD03839832
  • Cas Number : 3279-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-172
Synonym 6-Bromo-2-oxo-1,2,3,4-tetrahydroquinoline
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/6-bromo-34-dihydroquinolin-21h-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromo-34-dihydroquinolin-21h-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart