APO455830005 Apollo Scientific Ltd

4-Chloro-2-iodophenol

  • MDL number MFCD08235248
  • Cas Number : 71643-66-8
  • Molecular Weight : 254.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 74-76
Iupac Name 4-chloro-2-iodophenol
Inchi Code 1S/C6H4ClIO/c7-4-1-2-6(9)5(8)3-4/h1-3,9H
Inchi Key JKPLMQJLGBBFLO-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 74-76
Product Link https://store.apolloscientific.co.uk/product/4-chloro-2-iodophenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-2-iodophenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart