APO455830010 Apollo Scientific Ltd

Methyl 2-iodo-6-methylbenzoate

  • MDL number MFCD08061917
  • Cas Number : 103440-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 6-iodo-o-toluate, 3-Iodo-2-(methoxycarbonyl)toluene
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54871

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54871
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart