APO455830014 Apollo Scientific Ltd

1H-Indazole-5-sulphonyl chloride

  • MDL number MFCD11650650
  • Cas Number : 599183-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-(Chlorosulphonyl)-1H-indazole
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/1h-indazole-5-sulphonyl-chloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/1h-indazole-5-sulphonyl-chloride
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart