APO455830016 Apollo Scientific Ltd

3-Methylisatoic anhydride

  • MDL number MFCD03426399
  • Cas Number : 66176-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 8-Methyl-2H-3,1-benzoxazine-2,4(1H)-dione, 8-Methyl-1H-benzo[d][1,3]oxazine-2,4-dione
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-methylisatoic-anhydride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-methylisatoic-anhydride
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart