APO455830017 Apollo Scientific Ltd

3-Methyl-4-nitrobenzaldehyde

  • MDL number MFCD08445621
  • Cas Number : 18515-67-8
  • Molecular Weight : 165.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-methyl-4-nitrobenzaldehyde
Inchi Code 1S/C8H7NO3/c1-6-4-7(5-10)2-3-8(6)9(11)12/h2-5H,1H3
Inchi Key WHJDQIWWIJARMK-UHFFFAOYSA-N

Description

Synonym 5-Formyl-2-nitrotoluene, 4-Formyl-2-methylnitrobenzene
Purity 97%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-methyl-4-nitrobenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-methyl-4-nitrobenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart