APO455830026 Apollo Scientific Ltd

2-Chloropyrimidine-4-carbonitrile

  • MDL number MFCD00234989
  • Cas Number : 75833-38-4
  • Molecular Weight : 139.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 53-55
Iupac Name 2-chloro-4-pyrimidinecarbonitrile
Inchi Code 1S/C5H2ClN3/c6-5-8-2-1-4(3-7)9-5/h1-2H
Inchi Key HXVQPZSXXYOZMP-UHFFFAOYSA-N

Description

Synonym 2-Chloro-4-cyanopyrimidine, 2-Chloro-4-cyano-1,3-diazine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 53-55
Product Link https://store.apolloscientific.co.uk/product/2-chloropyrimidine-4-carbonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloropyrimidine-4-carbonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart