APO455830027 Apollo Scientific Ltd

2-Chloropyrimidine-4-carboxamide

  • MDL number MFCD00234053
  • Cas Number : 22536-66-9
  • Molecular Weight : 157.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-4-pyrimidinecarboxamide
Inchi Code 1S/C5H4ClN3O/c6-5-8-2-1-3(9-5)4(7)10/h1-2H,(H2,7,10)
Inchi Key UXXQEVFRPLIOHJ-UHFFFAOYSA-N

Description

Synonym 4-(Aminocarbonyl)-2-chloropyrimidine, 4-Carbamoyl-2-chloropyrimidine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloropyrimidine-4-carboxamide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloropyrimidine-4-carboxamide
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart