APO455830029 Apollo Scientific Ltd

Cyclohexylacetaldehyde

  • MDL number MFCD02261737
  • Cas Number : 5664-21-1
  • Molecular Weight : 126.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 62
Iupac Name cyclohexylacetaldehyde
density 0.919
Inchi Code 1S/C8H14O/c9-7-6-8-4-2-1-3-5-8/h7-8H,1-6H2
Inchi Key JJMDTERTPNYIGZ-UHFFFAOYSA-N

Description

Melting Point 62
Synonym (2-Oxoethyl)cyclohexane
Purity 90%
Density 0.919
Flash Point 58-60/10mm
Storage temp. Store at -20
Physical Form Liquid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/cyclohexylacetaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/cyclohexylacetaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart