APO455830038 Apollo Scientific Ltd

Isoquinoline-8-carboxaldehyde

  • MDL number MFCD06227440
  • Cas Number : 787615-01-4
  • Molecular Weight : 157.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 8-Formylisoquinoline, 8-Formyl-2-azanaphthalene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/isoquinoline-8-carboxaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/isoquinoline-8-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart