APO455830041 Apollo Scientific Ltd

8-(Hydroxymethyl)isoquinoline

  • MDL number MFCD11869736
  • Cas Number : 1159511-15-5
  • Molecular Weight : 159.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-isoquinolinylmethanol
Inchi Code 1S/C10H9NO/c12-7-9-3-1-2-8-4-5-11-6-10(8)9/h1-6,12H,7H2
Inchi Key UDNDYRZGZAWJHB-UHFFFAOYSA-N

Description

Synonym (Isoquinolin-8-yl)methanol, 8-(Hydroxymethyl)-2-azanaphthalene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/8-hydroxymethylisoquinoline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/8-hydroxymethylisoquinoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart