APO455830044 Apollo Scientific Ltd

8-(Bromomethyl)isoquinoline hydrobromide

  • MDL number MFCD12674806
  • Cas Number : 1215541-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 8-(Bromomethyl)-8-azanaphthalene hydrobromide
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=44658

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=44658
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart