APO455830045 Apollo Scientific Ltd

Thieno[3,2-c]pyridine

  • MDL number MFCD01760493
  • Cas Number : 272-14-0
  • Molecular Weight : 135.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name thieno[3,2-c]pyridine
Inchi Code 1S/C7H5NS/c1-3-8-5-6-2-4-9-7(1)6/h1-5H
Inchi Key MKYRMMMSZSVIGD-UHFFFAOYSA-N

Description

Synonym 5-Aza-1-benzothiophene, 5-Azabenzo[b]thiophene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/thieno32-cpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/thieno32-cpyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart