APO455830053 Apollo Scientific Ltd

2,4-Dichloro-6-methylphenol

  • MDL number MFCD00019988
  • Cas Number : 1570-65-6
  • Molecular Weight : 177.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 53-56
Iupac Name 2,4-dichloro-6-methylphenol
Inchi Code 1S/C7H6Cl2O/c1-4-2-5(8)3-6(9)7(4)10/h2-3,10H,1H3
Inchi Key WJQZZLQMLJPKQH-UHFFFAOYSA-N

Description

Synonym 4,6-Dichloro-o-cresol, 3,5-Dichloro-2-hydroxytoluene
Flash Point 225-226
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 53-56
Product Link https://store.apolloscientific.co.uk/product/24-dichloro-6-methylphenol

Safety Information

signalword Danger
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/24-dichloro-6-methylphenol
pictograms GHS07;GHS05
hstatements H302;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart