APO455830057 Apollo Scientific Ltd

4-Bromo-2-methylphenol

  • MDL number MFCD00055435
  • Cas Number : 2362-12-1
  • Molecular Weight : 187.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62-67
Synonym 5-Bromo-2-hydroxytoluene, 4-Bromo-o-cresol
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-methylphenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-methylphenol
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart